Skin care ingredients
Novel, high-purity silicone ingredients, including surfactant-free silicone emulsion technology, silicone elastomer gels, gum blends and specialty fluids
From novel, patented, surfactant-free, silicone emulsion technology to gel delivery systems, NuSil® skin care ingredients from Avantor® create unique sensory profiles for skin care formulations. Our portfolio of high-purity silicone ingredients provides endless possibilities for beauty care manufacturers with encapsulation and suspension technology platforms, elastomer gels, gums and fluids available in a variety of diluents, viscosities and refractive indices.
- Surfactant-free silicone emulsion technology: Discover our breakthrough surfactant-free silicone emulsion technology, allowing silicone and water to coexist without the use of surfactants. The platform offers a unique dual-phase, transformable texture, letting you have the best of both worlds – a fresh and watery sensation followed by the substantial feel from the silicone.
- Elastomer gels: With the highest purity on the market, our elastomer gels bring a no-pilling, no-balling, luxurious and ultra-mattifying powdery feel to skin care and color cosmetics.
- Gum blends: Our film-forming, semi-occlusive gum blends offer a rich and powdery after-feel with increased shine.
- Specialty fluids: NuSil specialty fluids are transfer- and wash-off-resistant compared to standard dimethicone ingredients. They reduce tackiness in formulations and act as a hydrophobic and oleophobic ingredient to deliver moisturizing properties.
Custom formulations tailored to your needs
NuSil has the expertise, processes, and proprietary technology to customize our silicones based on your unique specifications. We leverage our insight into the unique silicone chemistry and material properties to supply off-the-shelf and custom formulations tailored to your process and end-application performance requirements.
Learn more about how we can help you get the products you need.
Product finder
Skin care
NuSil silicone ingredients have offered unrivaled purity and innovative technology. Skin care industry leaders rely on NuSil for innovative textures with the lighter, fresher feel consumers demand in skin care and color cosmetics, from refreshing serums to night masks.
Related content
Browse additional resources related to innovative silicone skin care ingredients.
Customer support
Contact us
Ready to move your next innovation forward? NuSil can help.
How can we help you?
NuSil experts will help you find the right silicone skin care ingredient for your application, whether you need formulations using surfactant-free silicone emulsion technology or a specialty fluid.
© 2023 Avantor, Inc. | Terms of use | Privacy policy | EEO and Affirmative Action Statement | Sitemap
© 2023 FORTUNE Media IP Limited All rights reserved. Used under license.