Adhesives and Sealants
NuSil® high-purity silicone adhesives and sealants from Avantor® provide strong bonds for the most demanding applications, including medical devices and space craft.
Industry-leading silicone adhesives and sealants
Our line of NuSil® high-performance silicone adhesives and sealants are formulated to meet your needs, whether for biocompatible silicone adhesives, medical-grade silicones, or sealants for use in extreme temperatures.
We work with you to help you find the correct product by balancing all your needs, such as manufacturing throughput, device performance, reliability, and durability. Whether you need a biocompatible or ultra-low outgassing silicone or something in between, our experts will guide you to the best choice.
NuSil offers silicone adhesives to boost manufacturing throughout and increase performance:
- One-part room temperature vulcanized (RTV) silicone adhesives: Our one-part moisture cure formulations do not require mixing prior to use. They are ideal for use with temperature-sensitive components.
- Two-part RTV, or heat-curing silicone adhesives: These platinum-catalyzed formulations rapidly cure without byproducts or leaving groups. Two-part RTV silicones are ideal for use where you want faster throughput.
- Dispersed silicone adhesives: NuSil dispersed adhesives create a thin, uniform layer. Their low viscosity lets them easily flow into hard-to-reach areas.
- Film adhesives & tapes: Our double-sided tapes and film adhesives are an alternative to traditional liquid adhesives. They are available in sheet form with release liners and an internal structural support layer. These adhesives offer reliable bond line control in a peel-and-stick format.
- Pressure-sensitive adhesives (PSAs): This solvent-based, removable adhesive is non-curing but forms a strong bond when pressure is applied.
- Soft silicone adhesives (SSAs): Our SSAs provide permeability, protection, and atraumatic removal from human skin. They are ideal for wound care, scar management, and wearable monitoring devices.
Our silicone portfolio also includes primers to enhance adhesion.
Custom formulations tailored to your needs
NuSil has the expertise, processes, and proprietary technology to customize our silicones based on your unique specifications. We leverage our insight into the unique silicone chemistry and material properties to supply off-the-shelf and custom formulations tailored to your process and end-application performance requirements.
Learn more about how we can help you get the products you need.
Product Finder: Find NuSil silicone adhesives & sealants for your application
Medical implants
Manufacturers rely on NuSil silicone adhesives for a wide range of medical device applications, from cochlear implants and pacemakers to gastric balloons. Our adhesives for long-term implantable devices (>29 days) help create efficiencies in the production process by reducing manufacturing time and improving device performance.
General healthcare
Biocompatible silicone adhesives from NuSil are ideal for assembling medical devices for short-term use in the body (<29 days) or for external applications such as wound care. Our off-the-shelf and customized products are supplied with the regulatory support needed to bring your product to market.
Space
NuSil low and ultra-low outgassing adhesives are specifically designed to increase manufacturing throughput while reliably binding to many substrates, including metals, plastics, glass and polyimide. Our industry-leading, space-grade tapes and film adhesives offer less touch time and faster cure.
Aviation and defense
From metals to glass to plastics, NuSil silicone solutions create a reliable bond suitable for mission-critical applications in commercial aviation and defense applications. We offer off-the-shelf and custom formulations to meet specific requirements, such as size, thickness or adhesion to specific substrates.
Optoelectronics and electronics
We offer an extensive range of optically clear, reflective index (RI) silicone matching adhesives. NuSil advanced technologies can help manufacturers achieve improved device performance with longer operating lives.
Additional information about silicone adhesives.
NuSil® by Avantor is constantly adding to its comprehensive library of resources covering its complete line of high-purity silicone adhesives and sealants.
Durometer
Tensile, tear & elongation
Modulus
Viscosity
Extrusion rate
Healthcare
Silicone adhesives
We offer a wide range of NuSil® brand silicone adhesives and silicone primers to improve the productivity of medical device manufacturing. See the difference we can make.
Customer support
Contact us
Ready to move your next innovation forward? NuSil can help.
How can we help you?
From medical adhesives for wound care to high-temperature silicone adhesive products, NuSil experts will help you find the right product for your application.
© 2023 Avantor, Inc. | Terms of use | Privacy policy | EEO and Affirmative Action Statement | Sitemap
© 2023 FORTUNE Media IP Limited All rights reserved. Used under license.