Silicone primers
NuSil® supplies expertly formulated silicone primers to improve the bond between silicone adhesives and the substrate being joined.
NuSil silicone primers help to ensure the effectiveness and long-lasting performance of silicone adhesives by preparing a substrate surface for adhesive application. They also aid in adhering to materials with low surface energy by activating the surface to provide the best bond possible.
The NuSil portfolio of silicone primers has been developed to help improve adhesion for a wide range of substrates: silicone, metals such as aluminum, stainless steel and titanium and a wide range of plastics and rubbers, including polytetrafluoroethylene, polycarbonate, polyurethane, polyimide, ABS, PEEK and polysulphone.
Our extensive line of silicone primers is moisture-sensitive and time-dependent: they will react to moisture in the air, and it is typically recommended that they cure for 10-15 minutes before a silicone adhesive is applied. They can be applied through a variety of methods including spraying, wiping, brushing or dipping.
NuSil can collaborate with you to assess the surface properties of your substrate and recommend both the best silicone adhesive and best silicone primer to work with your production processes and the devices you are manufacturing.
Custom formulations tailored to your needs
NuSil has the expertise, processes, and proprietary technology to customize our silicones based on your unique specifications. We leverage our insight into the unique silicone chemistry and material properties to supply off-the-shelf and custom formulations tailored to your process and end-application performance requirements.
Learn more about how we can help you get the products you need.
Product finder
Medical implants
Medical device manufacturers joining parts with silicone adhesives can count on NuSil silicone primers to help create the best bond possible.
General healthcare
When assembling devices used in general healthcare applications, NuSil silicone primers help produce components with long-lasting bonds.
Space
NuSil offers silicone primers specially formulated for elements of spacecraft including solar panels and other electronic devices that will face the harsh conditions of outer space.
Aviation & defense
NuSil offers a wide range of primers specifically formulated for use with substrates commonly used on aviation and defense platforms.
Optoelectronics & electronics
From adhesion to potting and encapsulating, primers ensure the best bond between the silicone adhesive and underlying substrates.
Related content
Browse additional resources related to silicone primers.
Advanced technologies
Healthcare
Advanced technologies
Customer support
Contact us
Ready to move your next innovation forward? NuSil can help.
How can we help you?
NuSil experts will help to ensure that you choose the right NuSil silicone primer based on the adhesive you are using, and the substrates being adhered.
© 2023 Avantor, Inc. | Terms of use | Privacy policy | EEO and Affirmative Action Statement | Sitemap
© 2023 FORTUNE Media IP Limited All rights reserved. Used under license.