Coatings & dispersions
Silicone coatings and dispersions for protection in power electronics and biocompatibility in medical devices
NuSil® coatings and dispersions from Avantor® create and protect critical parts or technologies for a wide range of applications in the healthcare, space, aviation, defense, optoelectronics and electronics industries.
NuSil silicone coatings and dispersions are elastomers dispersed in a solvent for ease of application. They can be applied in very thin layers to evenly coat a device or surface or used to create a film or thin-walled part with strong elastomeric properties.
NuSil’s broad range of silicone coatings and dispersions offer numerous options to accommodate processing requirements and functional properties:
- One-part or two-part coatings: Our coatings are available as a one-part formulation with no mixing required or two-part configuration offering a longer work time.
- Solvent-based or solventless coatings: Solvent-based coatings are ideal for processes where the silicone is applied as a thin film coating. Solvent-less coatings are non-flammable and do not require controlled environments for handling.
- Special features: NuSil coating options include hydrocarbon-resistance, lubrication, low coefficient of friction and high clarity formulations.
Custom formulations tailored to your needs
NuSil has the expertise, processes, and proprietary technology to customize our silicones based on your unique specifications. We leverage our insight into the unique silicone chemistry and material properties to supply off-the-shelf and custom formulations tailored to your process and end-application performance requirements.
Learn more about how we can help you get the products you need.
Product finder
Medical implants
For long term implanted devices (>29 days) NuSil silicone coatings and dispersions are ideal for dipping and spraying processes that produce thin elastomeric films or forms.
General healthcare
Our portfolio of medical-grade coatings and dispersions offer solutions for devices implanted in the body for < 29 days, such as balloons or external devices like needles.
Space
Our space-grade coatings protect against atomic oxygen (AO) while providing resistance to radiation and are used on a variety of spacecraft surfaces and small electronic components.
Aviation & defense
NuSil coatings feature fuel and heat-resistant properties that protect critical technology in missiles, rockets, launch equipment and unmanned aerial vehicles.
Optoelectronics & electronics
From extending the operating life of circuit boards to providing stability for elastomers, our conformal coatings protect sensitive elements from moisture, contaminants, shock and temperature extremes.
Related content
Browse additional resources related to silicone conformal coatings.
Healthcare
Advanced technologies
Advanced technologies
Advanced technologies
Healthcare
Advanced technologies
Durometer
Tensile, tear & elongation
Modulus
Viscosity
Conformal coatings
The NuSil® brand of silicone conformal coatings efficiently and reliably protects electronic components and helps ensure long operating life. Let us put our silicone expertise to work for you.
Customer support
Contact us
Ready to move your next innovation forward? NuSil can help.
How can we help you?
Our experienced team will identify or develop the best solution for your application, whether you need a sprayable silicone dispersion or a heat-resistant coating technology.
© 2023 Avantor, Inc. | Terms of use | Privacy policy | EEO and Affirmative Action Statement | Sitemap
© 2023 FORTUNE Media IP Limited All rights reserved. Used under license.