R-1505
NON-CORROSIVE STATIC DISSIPATIVE RTV SILICONE
Description
  • One-part, black, electrically conductive and static dissipative RTV silicone
  • Cures at room temperature to form a flexible, resilient, conductive bond or seal
  • Good adhesion across a broad temperature range
  • Based on a dimethyl diphenyl silicone copolymer
  • Oxime cure system
Applications
  • For applications requiring a non-corrosive adhesive / sealant
  • For RFI and EMI shielding for industrial and electronic applications
  • Provides protection from extremes in temperature, humidity, radiation, thermal stress and mechanical stress
  • Excellent for form-in-place conductive gaskets and for bleeding off static electricity
  • Provides good radiation resistance
  • For applications requiring an operating temperature range of -115°C to 260°C (-175°F to 500°F)
Product Details
Property Average Result
Volume Resistivity 8 ohmcm
Cure System Oxime
Durometer 75 Type A
Tensile 525 psi (3.62 MPa)
Appearance Black
Cure 7 days / 25 °C
Dielectric Strength 10 volts/mil (0.39 kV/mm)
Elongation 25 %
Flow Rate 0.1 in (2.54 mm)
Lap Shear 290 psi (2 MPa)
Mix Ratio 1 PART
Non Volatile Content 100 %
Specific Gravity 1.23
Tack Free Time 10 minutes
Thermal Conductivity 0.3 W/(mK)
Comment 8 ohmcm. Static Dissipation