Fluorosilicones for Aviation and Defense Applications – Fuel Resistance
Our NuSil® brand of fluorosilicones has an established track record of providing resistance from degradation during prolonged fuel and solvent exposure.
Decrease aircraft downtime in aerospace industry where failure is not an option
Fuel and common solvents on aircraft can degrade materials, leading to additional maintenance, unplanned downtime or catastrophic failure. This unique challenge leads aviation and defense manufacturers to often choose fluorosilicones for their engineered resistance to degradation from exposure to hydrocarbons and temperature extremes.
NuSil sets the standard for fluorosilicone performance and purity
- Formulated to maintain dimensional shape, integrity, and function when exposed to hydrocarbons for superior stability compared to other materials
- Broad portfolio of custom and standard solutions, including adhesives, sealants, coatings, sheeting, molding materials, gels and foams
- Strong regulatory support and proven history: AS9100 and ISO 9001 certified; compliant with applicable requirements; and expertise built on a flight heritage of more than 40 years.
Making the right choice for fuel- and solvent-resistant fluorosilicones
There are many options when using a fluorosilicone as there are many forms and cure chemistries available. This versatility makes fluorosilicones ideal for diverse applications such as O-rings, gaskets, seals, molded parts, coatings, gap fillers, adhesives, sealants, potting and encapsulation. Alongside standard products, NuSil silicones can be customized for specific requirements.
Understanding the needs and constraints of an application can help clarify which fluorosilicone option is best. Here are some considerations when selecting the right fluorosilicone for your application:
High performance material for extreme conditions and specialized applications
Designed for mission-critical applications, fluorosilicones provide superior resistance to harsh conditions, including extreme temperatures and high altitudes, owing to their low volatility. Their ability to be customized with functional fillers enables specific electrical properties crucial for aerospace and defense systems. Furthermore, fluorosilicones can be color-matched to meet federal standards (FED-STD-595), ensuring consistency in regulated environments where uniformity is essential.
Download content
Browse additional resources related to fluorosilicones.
Find Fluorosilicone Products
Use our product search to find the right fluorosilicone solution for your application.
Customer support
Contact us
Ready to move your next innovation forward? NuSil can help.
The Industry Leader in Fluorosilicones for Aviation and Defense
NuSil silicones have been trusted for over 40 years for use in the most demanding applications. Our best-in-class product lines are the premier choice across the aviation and defense industries. We offer a wide range of customization and product development services to meet specific application requirements to help ensure your vital equipment fulfills its mission.
© 2023 Avantor, Inc. | Terms of use | Privacy policy | EEO and Affirmative Action Statement | Sitemap
© 2023 FORTUNE Media IP Limited All rights reserved. Used under license.