XL-151
PENDENT HYDRIDE CROSSLINKER
Description
- Pendent methylhydrosiloxane - dimethylsiloxane co-polymer crosslinker
- CAS # 68037-59-2
Applications
- For crosslinking silicone addition cure polymer systems, to form adhesives, sealant, rubbers and gels due to its reactivity with vinyl functional groups
Product Details
Property | Average Result |
---|---|
Refractive Index | 1.40 |
Viscosity | 480 cSt |
Mmol | 0.4 H/g |
% Pt | |
Appearance | Translucent |
Cure System | Non-Curing |
Mix Ratio | 1 PART |
Volatile Content | 0.2 % |