Description
- 2,4,6,8 –Tertramethyl-2,4,6,8-tetravinylcyclotetrasiloxane (95% minimum)
- CAS # 2254-06-5
Applications
- Competitive Inhibitor for adjusting cure rate of vinyl and hydride functional silicone systems that cure via hydrosilylation
- Compatible with a variety of silicone polymer systems for formulating silicone adhesives, elastomers and gels due to its reactivity and solubility with silicone
Product Details
Property | Average Result |
---|---|
Appearance | Clear |
Cure System | Non-Curing |
Mix Ratio | 1 PART |
Mmol | 11.5 Vi/g |