Description
- Specially formulated primer designed for use with platinum-cured systems where conventional silicone primers are insufficient
- One-component primer supplied in VM&P Naphtha requires no mixing
- Air-drying
- Convenient container sizes produce less waste
Applications
- Improves the adhesion of addition-cured systems to iridite coated materials as well as other various substrates including: metals, glass, ceramics, some plastics, and other silicone material
Product Details
Property | Average Result |
---|---|
Non Volatile Content | 15 % |
Specific Gravity | 0.8 |
Appearance | Translucent |
Cure System | Condensation |
Lap Shear | 166 psi (1.14 MPa) |
Refractive Index | 1.40 |
Comment | Improved adhesion to gold - Recommended where platinum inhibition is of concern such as when bonding on polyphenylsulphone. |