SP-121
SILICONE PRIMER
Description
  • Specially formulated primer designed for use with one and two-part RTV systems where conventional silicone primers are insufficient
  • Red in color to provide easy visual determination of surface coverage
  • One-component primers supplied in VM&P Naphtha require no mixing
  • Air-drying
  • Convenient container sizes produce less waste
Applications
  • Improves the adhesion of one and two-part RTV systems to various substrates including: metals, glass, ceramics, rigid plastics, and wood
Product Details
Property Average Result
Non Volatile Content 3.2 %
Specific Gravity 0.76
Appearance Red
Cure System Condensation
Lap Shear 300 psi (2.07 MPa)
Comment Use with addition or condensation cure.