Description
- Two-part, black electrically conductive RTV silicone
- 10:1 Mix Ratio (Part A:B)
Applications
- For applications requiring Ultra Low OutgassingTM and minimal volatile condensables under extreme operating conditions to avoid condensation in sensitive devices
- Use for RFI and EMI shielding in electronic and space applications
- Use to adhere covers onto housings or for any application where grooves and other configurations require a flowable to limited flow material
- For applications requiring a broader operating temperature range
Product Details
Property | Average Result |
---|---|
Volume Resistivity | 2.5 ohmcm |
Cure System | Platinum |
Durometer | 75 Type A |
Tensile | 475 psi (3.28 MPa) |
Appearance | Black |
Cure | 30 minutes / 150 °C |
Elongation | 90 % |
Lap Shear | 250 psi (1.72 MPa) |
Lap Shear | 250 psi (1.72 MPa) |
Mix Ratio | 10:1 |
Special Feature | |
Specific Gravity | 1.19 |
Work Time | 2 hours |