Description
- Two-part, optically clear silicone system
- 10:1 Mix Ratio (Part A:B)
Applications
- For electronic and space applications requiring Ultra Low OutgassingTM and minimal volatile condensables to avoid condensation in sensitive devices
- As an embedding or potting compound for environmental protection of electronic assemblies and components
- Provides protection from humidity, radiation, thermal stress and mechanical stress
- As an adhesive in low-strength applications such as solar cell arrays where clarity and low volatility are important
- For applications requiring a broader operating temperature range
Product Details
Property | Average Result |
---|---|
Cure System | Platinum |
Viscosity | 9,500 cP (9,500 mPa*s) |
Lap Shear | 375 psi (2.59 MPa) |
Durometer | 45 Type A |
Appearance | Clear |
Cure | 15 minutes / 150 °C |
Elongation | 125 % |
Mix Ratio | 10:1 |
Special Feature | |
Specific Gravity | 1.02 |
Tensile | 950 psi (6.55 MPa) |
Comment | Pourable |