Description
- Two-part, red, low specific gravity, RTV silicone
- 1:1 Mix Ratio (Part A: Part B)
Applications
- For applications requiring Ultra Low OutgassingTM and minimal volatile condensables under extreme operating conditions to avoid condensation in sensitive devices
- Use a bonding, sealing, or potting material in electronic and space applications
- Provides radiation resistance, low thermal conductivity, oxidation stability, thermal stability and good ablative characteristics
- Especially useful to bond solar cells to solar array panels
- For applications requiring a broader operating temperature range
Product Details
Property | Average Result |
---|---|
Cure System | Platinum |
Viscosity | 325,000 cP (325,000 mPa*s) |
Lap Shear | 175 psi (1.21 MPa) |
Durometer | 45 Type A |
Appearance | Red |
Cure | 30 minutes / 150 °C |
Elongation | 150 % |
Mix Ratio | 1:1 |
Special Feature | |
Specific Gravity | 0.74 |
Tensile | 225 psi (1.55 MPa) |
Work Time | 4 hours |
Comment | Fast Cure, Low Density, Primed Lap Shear 175psi |