Description
- Two-part, translucent silicone system
- Offers a high tear strength, good physical properties and a broad operating temperature range
- Convenient 1:1 mix ratio (Part A:Part B)
Applications
- For electronic and space applications requiring Ultra Low OutgassingTM and minimal volatile condensables under extreme operating conditions
- To provide protection of electronic components and assemblies against shock, vibration, moisture, dust, chemicals and other environmental hazards
- Ideal for use in potting connectors, cable harness breakouts, molded high-voltage terminals, seals and gaskets due to its high tear strength
- For applications requiring a broader operating temperature range
Product Details
Property | Average Result |
---|---|
Cure System | Platinum |
Viscosity | 77,000 cP (77,000 mPa*s) |
Lap Shear | 475 psi (3.28 MPa) |
Durometer | 35 Type A |
Appearance | Translucent |
Cure | 15 minutes / 150 °C |
Elongation | 300 % |
Mix Ratio | 1:1 |
Specific Gravity | 1.11 |
Tack Free Time | 7 hours |
Tear | 40 ppi (7.05 kN/m) |
Tensile | 700 psi (4.83 MPa) |
Work Time | 1 hours |
Comment | Ultra low Outgassing. Recommended for broad temperature applications. 475 psi (3.3 MPa), primed (CF1-135) lap shear. |