S-7400
HIGH/LOW TEMPERATURE SILICONE FLUID
Description
  • A clear dimethyl, diphenyl polysiloxane fluid
  • Available in viscosities from 40,000 to 2.5 million centistokes
  • Diphenyl content offers improved thermal stability
Applications
  • For maximum vibration control and damping in dash pots for time delay relays, pulsation dampers and meter movements
  • Use as the working media in fluid clutch and fluid drive systems in the aerospace and automotive industry
  • Consider for use as a lubricant, coating, release agent and performance additive
  • Provides high and low temperature stability, oxidation resistance, very low vapor pressure, low surface temperature, high flash point and optimum radiation and oxidation resistance.
Product Details
Property Average Result
Substrate Glass / Metal / Plastic
Volatile Content 4 %
Viscosity 60,000 cP
Appearance Translucent
Dielectric Strength 400 volts/mil (15.6 kV/mm)
Operating Temperature -65 °C / 260 °C
Refractive Index 1.43
Specific Gravity 1.1
Volume Resistivity 1,000,000,000,000,000 ohmcm