Description
- One-part RTV silicone elastomer dispersed in tert butyl acetate
- Based on a fluorosilicone copolymer
Applications
- For coating, sealing and bonding applications requiring solvent and/or fuel resistance
- Designed for spraying, dip casting, and RTV curing of thin, elastomeric films
- For use as a coating to minimize ice adhesion
- Bonds aggressively to most surfaces
- Slight addition of heat will accelerate cure
Product Details
Property | Average Result |
---|---|
Viscosity | 1,625 cP (1,625 mPa*s) |
Non Volatile Content | 60 % |
Appearance | Translucent |
Attribute | |
Cure | 72 hours / RTV |
Cure System | Acetoxy |
Durometer | 25 Type A |
Elongation | 400 % |
Mix Ratio | 1 PART |
Specific Gravity | 1.29 |
Tear | 35 ppi (6.17 kN/m) |
Tensile | 425 psi (2.93 MPa) |
Comment | Dispersion Coating, 60% Solids |