R-2682-12
RTV SILICONE FILM ADHESIVE
Description
  • Two-part film adhesive
  • Cures at room temperature or rapidly with heat
Applications
  • For bonding or sealing silicone elastomers and some metals or plastics
  • For applications requiring shorter processing times, easy clean-up, and consistent bond thickness
Product Details
Property Average Result
Cure System Platinum
Lap Shear 100 psi (0.69 MPa)
Appearance Translucent
Application Life 4 minutes
Cure 4 hours / 65 °C
Comment For applications requiring shorter processing times, easy clean-up, and consistent bond thickness. Adheres to silicone, and some metals or plastics. Sheet is 0.013 inches (0.3 mm) and cures on surfaces where the Activator is applied. Cures at room temperature or rapidly with heat.