Description
- Two-part, tan, electrically conductive RTV silicone
- Based on a methyl silicone polymer with a broad temperature range
- Designed to cure at room temperature and at elevated temperatures
- 20:1 Mix Ratio (Part A:Part B)
Applications
- For RF and EMI shielding for electrical and aerospace applications
- Excellent for formed-in-place conductive gaskets and grounding connections
- Use to adhere covers on housings or where grooves or other configurations require a non-flowable to limited flow material
Product Details
Property | Average Result |
---|---|
Volume Resistivity | 0.01 ohmcm |
Cure System | Platinum |
Durometer | 60 Type A |
Tensile | 210 psi (1.45 MPa) |
Appearance | Tan |
Cure | 30 minutes / 150 °C |
Dielectric Strength | 5 volts/mil (0.2 kV/mm) |
Elongation | 275 % |
Mix Ratio | 20:1 |
Specific Gravity | 3.6 |
Tack Free Time | 5 hours |
Thermal Conductivity | 1.5 W/(mK) |
Work Time | 4 hours |
Comment | Electrically Conductive RTV Silicone. Mixes to smooth paste, can be screen printed. |