Description
- Two-part, RTV silicone foam
- Yields a low density silicone rubber foam when catalyzed
- 100:6 Mix Ratio (Base: Catalyst)
Applications
- Use in shock and vibration dampening situations requiring a lightweight, flexible foam with excellent thermal and electrical properties
Product Details
Property | Average Result |
---|---|
Viscosity | 4,700 cP (4,700 mPa*s) |
Cure System | Tin |
Foam Density | 10 lbs/ft3 (0.16 g/cm3) |
Application Life | 28 minutes |
Appearance | Tan |
Cure | 10 minutes / RTV |
Mix Ratio | 100:6 |
Non Volatile Content | 100 % |
Comment | Low Density / Soft, 10 lb/ft^3 (0.16 g/cm^3) |