Description
- Two-part, room temperature curing, silicone foam
- Platinum cure
- No odor
- Yields a low density silicone foam when catalyzed
- 1:1 Mix Ratio (Part A: Part B)
Applications
- In shock and vibration dampening situations requiring a light weight, flexible foam with excellent thermal insulation.
- Applications requiring a low density component for better fuel economy, and other applications where the weight needs to be considered.
Product Details
Property | Average Result |
---|---|
Viscosity | 40,000 cP (40,000 mPa*s) |
Cure System | Platinum |
Foam Density | 12 lbs/ft3 (0.19 g/cm3) |
Application Life | 2 minutes |
Appearance | White |
Cure | 20 minutes / RTV |
Mix Ratio | 1:1 |
Comment | Flexible cure in place low density foam. Application life 2 minutes. |