R-2180-2
ADDITION-CURE HIGH TEAR STRENGTH, SILICONE DISPERSION
Description
  • Two-part silicone elastomer dispersed in xylene
  • Two medium viscosity parts blend easily in a convenient 1:1 ratio (Part A:B)
Applications
  • For dip casting and heat-curing of thin elastomeric films
Product Details
Property Average Result
Viscosity 3,275 cP (3,275 mPa*s)
Non Volatile Content 20 %
Appearance Translucent
Attribute
Cure 30 minutes / RTV 45 minutes / 75 °C 135 minutes / 150 °C
Cure System Platinum
Durometer 45 Type A
Elongation 1,000 %
Mix Ratio 1:1
Stress at Strain 140 psi / 100 % (0.97 MPa / 100 %)
Tear 300 ppi (52.91 kN/m)
Tensile 1,650 psi (11.38 MPa)
Comment Cure: 30 M / 25 C : 45 M / 75 C : 135 M / 150 C, 20% Solids