Description
- Two Part, Translucent, 1:1 Mix ratio (A:B)
- Cures at room temperature or rapidly with the application of heat in an oven, or by a heat gun or lamp
- Non-slump but is easily dispensed
- Does not require atmospheric moisture to cure
- No curing byproducts such as acetic acid or methyl alcohol
- REACh and RoHS compliant
Applications
- As adhesives for bonding and sealing silicones to each other and other substrates such as metals and plastics
- Consistency allows products to be supplied in easy-to-use, airless side-by-side kits that eliminate mixing and deairing difficulties
- Protects electrical components and assemblies against shock, vibration, moisture, dust, chemicals, and other environmental hazards
Product Details
Property | Average Result |
---|---|
Viscosity | 180,000 cP (180,000 mPa*s) |
Durometer | 40 Type A |
Appearance | Translucent |
Dielectric Strength | 630 volts/mil (24.57 kV/mm) |
Elongation | 250 % |
Flow Rate | 0.3 in (7.62 mm) |
Specific Gravity | 1.1 |
Tack Free Time | 4 hours |
Tensile | 650 psi (4.48 MPa) |
Volatile Content | 0.3 % |
Work Time | 1.5 hours |