Description
- One-part flowable, RTV silicone dispersed in xylene
- Cures at room temperature within 24 hours upon exposure to atmospheric moisture
- Non-corrosive oxime cure system
- Electrically insulating
Applications
- Conformal coating for rigid and flexible printed circuit boards (PCBs).
- Protects from moisture ingress, vibration, shock, dust, chemicals, and other environmental hazards
- Low viscosity ideal for use as sprayable or dip coatings
- Suitable for dip casting into thin films
Product Details
Property | Average Result |
---|---|
Viscosity | 1,300 cP (1,300 mPa*s) |
Non Volatile Content | 70 % |
Appearance | Translucent |
Dielectric Strength | 513 volts/mil (20.01 kV/mm) |
Durometer | 23 Type A |
Elongation | 223 % |
Tack Free Time | 50 minutes |
Tensile | 235 psi (1.62 MPa) |
Thermal Conductivity | 0.17 W/(mK) |