Implant Line

MED6-6606

RTV SILICONE DISPERSION

Description

  • One part, RTV silicone dispersion
  • Cures at room temperature upon exposure to atmospheric moisture

Applications

  • Suitable for dip casting and heat-curing of thin elastomeric films
  • For use as an adhesive for bonding and sealing silicone together
  • Low viscosity makes dispersions ideal for use as sprayable coatings
  • Provides good adhesion to metals and other substrates
  • Designed for dipping or wiping
NuSil Technology’s MED6-6606 may be considered for use in human implantation for a period of greater than 29 days.

Product Details

Property Average Result
Cure Type RTV
Mix Ratio 1 PART
Rheology Flowable / Self Leveling
Appearance Translucent
Cure 3 days / RTV
Cure System Acetoxy
Durometer 20 Type A
Elongation 800 %
Non Volatile Content 30 %
Specific Gravity 1.11
Stress @ Strain
Substrate Primerless: Aluminum / Glass / Polyurethane / Silicone / Titanium / With Primer: Polycarbonate / Stainless steel
Tear 115 ppi (20.28 kN/m)
Tensile 1,190 psi (8.21 MPa)
Viscosity 95 cP (95 mPa*s)
Comment Silicone adhesive dispersed in heptane

Encapsulation Technology

Watch our CareSil team explain the breakthrough Encapsulation Technology and demonstrate how easy it is to create transformative formulations with CES.

Learn more

The test data shown for this material is the average value for typical properties. All of these properties may not be tested on a lot to lot basis and cannot be used to draft specifications. Please contact NuSil for assistance and recommendations in establishing limits for product specifications.