Premium Care Line

MED50-4900-6

COLOR MASTERBATCH FOR LIQUID SILICONE ELASTOMERS

Description

  • A translucent green, single component masterbatch, with the consistency of a low viscosity paste
  • Consists of pigment dispersed in a vinyl-functional silicone polymer which covalently bonds into the matrix of platinum-cured silicone systems
  • Supported by USP Class VI and ISO 10993 Biological testing (reference Biological Testing Data Table)

Applications

  • For easy and more precise pigmentation of liquid silicone rubbers and low consistency elastomers suitable for injection molding
  • Reduces production time and eliminates use of powders that may contaminate clean room environments
  • Strict quality controls ensure color consistency
NuSil Technology's MED50-4900-6 shall not be considered for use in human implantation for a period of greater than 29 days.

Product Details

Property Average Result
Appearance Translucent Green
Mix Ratio 1 PART
Comment Masterbatch to add color to platinum cure systems

Encapsulation Technology

Watch our CareSil team explain the breakthrough Encapsulation Technology and demonstrate how easy it is to create transformative formulations with CES.

Learn more

The test data shown for this material is the average value for typical properties. All of these properties may not be tested on a lot to lot basis and cannot be used to draft specifications. Please contact NuSil for assistance and recommendations in establishing limits for product specifications.