Implant Line

MED3-4800

TITANIUM DIOXIDE HEALTHCARE SILICONE COLOR MASTERBATCH

Description

  • A white single component masterbatch, with the consistency of a low viscosity paste
  • Consists of pigment dispersed in a vinyl-functional silicone polymer which covalently bonds into the matrix of platinum-cured silicone systems

Applications

  • For easy and more precise pigmentation of liquid silicone rubbers and low consistency elastomers suitable for injection molding
  • Reduces production time and eliminates use of powders that may contaminate clean room environments
NuSil Technology’s MED3-4800 may be considered for use in human implantation for a period of greater than 29 days.

Product Details

Property Average Result
Appearance White
Mix Ratio 1 PART
Comment LSR Masterbatch, 50% TiO2

Encapsulation Technology

Watch our CareSil team explain the breakthrough Encapsulation Technology and demonstrate how easy it is to create transformative formulations with CES.

Learn more

The test data shown for this material is the average value for typical properties. All of these properties may not be tested on a lot to lot basis and cannot be used to draft specifications. Please contact NuSil for assistance and recommendations in establishing limits for product specifications.