Description
- A white, single component masterbatch, with the consistency of a low viscosity paste
- Consists of Barium Sulfate dispersed in a vinyl-functional silicone polymer which covalently bonds into the matrix of platinum-cured silicone systems
Applications
- For easy and more precise addition of Barium Sulfate to liquid silicone rubbers and low consistency elastomers suitable for injection molding
- Reduces production time and eliminates use of powders that may contaminate clean room environments
Product Details
Property | Average Result |
---|---|
Appearance | White |
Mix Ratio | 1 PART |
Comment | 50% BaSO4, Barium Sulfate LSR Masterbatch For Radio Opacity |
Encapsulation Technology
Watch our CareSil team explain the breakthrough Encapsulation Technology and demonstrate how easy it is to create transformative formulations with CES.
The test data shown for this material is the average value for typical properties. All of these properties may not be tested on a lot to lot basis and cannot be used to draft specifications. Please contact NuSil for assistance and recommendations in establishing limits for product specifications.