Description
- Two-part silicone elastomer dispersed in xylene
- Cures to a thin silicone film that decreases the coefficient of friction (CoF) compared to the uncoated silicone and increases abrasion resistance for moving, sliding, and rubbing parts
- Cures via addition-cure chemistry
- 1:1 Mix Ratio (Part A: Part B)
Applications
- Low viscosity makes dispersions ideal for use as sprayable coatings
- For coating of tubing (ID/OD), balloons, valves, stoppers, o-rings, and for other silicone parts
Product Details
Property | Average Result |
---|---|
Cure System | Platinum |
Non Volatile Content | 25 % |
Appearance | Translucent |
Cure | 5 minutes / RTV |
Mix Ratio | 1:1 |
Solvent | Xylene |

Encapsulation Technology
Watch our CareSil team explain the breakthrough Encapsulation Technology and demonstrate how easy it is to create transformative formulations with CES.
The test data shown for this material is the average value for typical properties. All of these properties may not be tested on a lot to lot basis and cannot be used to draft specifications. Please contact NuSil for assistance and recommendations in establishing limits for product specifications.