Premium Care Line
MED-9031
HIGH CONSISTENCY SILICONE GREASE
Description
  • A high consistency dimethyl silicone grease
  • Non-curing
  • Available in a variety of packaging options for convenient dispensing and application
Applications
  • Effective high temperature lubricant for gaskets, o-rings, and other sealing mechanisms
  • Ideal for use in high vacuum/pressure systems on glass and ceramic joints
NuSil Technology’s MED-9031 shall not be considered for use in human implantation for a period of greater than 29 days.
Product Details
Property Average Result
Volatile Content 0.3 %
Viscosity 900,000 cP (900,000 mPa*s)
Appearance White
Specific Gravity 1.11
Substrate Glass / Metal / Plastic
Comment High Consistency Grease