Description
- A medium consistency dimethyl silicone grease
- Non-curing
- Available in a variety of packaging options for convenient dispensing and application
Applications
- Effective high temperature lubricant for gaskets, o-rings, and other sealing mechanismsIdeal for use in high vacuum/pressure systems on glass and ceramic joints
Product Details
Property | Average Result |
---|---|
Volatile Content | 0.5 % |
Viscosity | 300,000 cP (300,000 mPa*s) |
Appearance | White |
Specific Gravity | 1.08 |
Substrate | Glass / Metal / Plastic |
Comment | Medium Consistency Grease |