Description
- A low consistency dimethyl silicone grease
- Non-curing
- Available in a variety of packaging options for convenient dispensing and application
Applications
- Effective high temperature lubricant for gaskets, o-rings, and other sealing mechanisms
- Ideal for use in high vacuum/pressure systems on glass and ceramic joints
Product Details
Property | Average Result |
---|---|
Volatile Content | 0.3 % |
Viscosity | 95,000 cP (95,000 mPa*s) |
Appearance | Translucent |
Non Volatile Content | 99.4 % |
Specific Gravity | 1 |
Substrate | Glass / Metal / Plastic |
Comment | Low Consistency Grease |