Description
- Two-part, pourable, optically clear, silicone elastomer
- Cures with heat via addition-cure chemistry
- 1:1 Mix Ratio (Part A: Part B)
Applications
- For use in potting, encapsulating, coating, and injection or transfer molding applications
Product Details
Property | Average Result |
---|---|
Durometer | 40 Type A |
Viscosity | 66,000 cP (66,000 mPa*s) |
Work Time | 4.5 hours |
Tensile | 670 psi (4.62 MPa) |
Appearance | Clear |
Cure | 30 minutes / 150 °C |
Cure System | Platinum |
Elongation | 210 % |
Mix Ratio | 1:1 |
Refractive Index | 1.43 |
Specific Gravity | 1.43 |
Tear | 40 ppi (7.05 kN/m) |
Comment | 1.43 R.I., Optically Clear |