Description
- A high consistency silicone grease containing dimethyl methyl-trifluoropropylsiloxane copolymer
- Non-curing
- Available in a variety of packaging options for convenient dispensing and application
Applications
- Effective high temperature lubricant for gaskets, o-rings, and other sealing mechanisms
- Ideal for use in high vacuum/pressure systems on glass and ceramic joints
- Useful in applications requiring solvent resistance
- For lubricating the surfaces of molded dimethyl silicone parts
Product Details
Property | Average Result |
---|---|
Volatile Content | 0.6 % |
Viscosity | 2,000,000 cP (2,000,000 mPa*s) |
Substrate | Silicone |
Comment | High Consistency Methyl Fluoro Grease |