Implant Line
MED-6670
LOW COEFFICIENT OF FRICTION SILICONE COATING
Description
  • Two-part silicone elastomer dispersed in xylene
  • Cures to a smooth silicone film that decreases the coefficient of friction (CoF) compared to the uncoated silicone and increases abrasion resistance
  • Cures via addition-cure chemistry
  • 1:1 Mix Ratio (Part A: Part B)
Applications
  • Suitable for dip casting and heat-curing of thin elastomeric films
  • Low viscosity makes dispersions ideal for use as sprayable coatings
  • To provide a heat-cured, low-friction coating on cured silicone substrates
  • For tubing (ID/OD). Balloons, valves, stoppers, o-rings, and for moving or sliding parts
NuSil™ MED-6670 may be considered for use in human implantation for a period of greater than 29 days.
Product Details
Property Average Result
Cure System Platinum
Non Volatile Content 25 %
Appearance Translucent
Cure 5 minutes / RTV
Mix Ratio 1:1
Special Feature
Specific Gravity 0.96
Work Time 24 hours
Comment Low Coefficient of Friction Coating