Description
- Two-part medium viscosity silicone elastomer dispersed in heptane
- Cures via addition-cure chemistry
- 1:1 Mix Ratio (Part A: Part B)
Applications
- Suitable for dip casting and heat-curing of thin elastomeric films
- Low viscosity makes dispersions ideal for use as sprayable coatings
- Useful as a solvent-based adhesive
Product Details
Property | Average Result |
---|---|
Cure Type | Heat Cure |
Mix Ratio | 1:1 |
Rheology | Flowable / Self Leveling |
Appearance | Translucent |
Cure | 30 minutes / RTV |
Cure System | Platinum |
Durometer | 72 Type 00 |
Elongation | 750 % |
Non Volatile Content | 32 % |
Stress @ Strain | |
Substrate | Primerless: Aluminum / Glass / PMMA / Silicone / With Primer: ABS / PEEK / Polycarbonate / Polyimide / Polysulphone / Polyetherimide / Polyurethane / Stainless steel |
Tear | 120 ppi (21.16 kN/m) |
Tensile | 1,060 psi (7.31 MPa) |
Viscosity | 65 cP (65 mPa*s) |
Comment | Silicone adhesive dispersed in heptane, cures rapidly with heat |