Description
- Two-part, addition –cure black silicone ink dispersed in xylene
- Cures via addition-cure chemistry
- 1:1 Mix Ratio (Part A: Part B)
Applications
- Ideal for use as a marking ink for silicone parts and other components on which the coating must maintain long-term stability
- Use in applications requiring a rapid cure schedule
- For use in pad and transfer printing applications
- Provides high opacity for single print applications
Product Details
Property | Average Result |
---|---|
Cure System | Platinum |
Viscosity | 800 cP (800 mPa*s) |
Non Volatile Content | 60 % |
Mix Ratio | 1:1 |
Appearance | Black |
Cure | 5 minutes / 150 °C |
Comment | Heat Curable Ink in Black |