Description
- One-part RTV silicone elastomer dispersed in xylene
- Cures at room temperature upon exposure to atmospheric moisture
Applications
- Suitable for dip casting and heat-curing of thin elastomeric films
- For use as an adhesive for bonding and sealing silicone together
- Low viscosity makes dispersions ideal for use as sprayable coatings
Product Details
Property | Average Result |
---|---|
Cure System | Acetoxy |
Viscosity | 700 cP (700 mPa*s) |
Non Volatile Content | 29 % |
Durometer | 25 Type A |
Appearance | Translucent |
Cure | 5 days / RTV |
Elongation | 980 % |
Mix Ratio | 1 PART |
Refractive Index | 1.40 |
Specific Gravity | 1.09 |
Stress at Strain | 110 psi / 300 % (0.76 Mpa / 300 %) |
Tear | 120 ppi (21.16 kN/m) |
Tensile | 1,500 psi (10.34 MPa) |
Comment | One Part, in Xylene |