Implant Line
MED-6601
ADDITION CURE SILICONE ELASTOMER SYSTEM
Description
  • Cures with heat via addition-cure chemistry
  • Intended to be dispersed in solvent at a suggested mix ratio of 100parts Part A: 1part Part B: 1part Part C: 234parts Xylene (or other suitable solvent)
Applications
  • Once dispersed, the material is suitable for dip casting and heat-curing of thin elastomeric films
NuSil Technology’s MED-6601 may be considered for use in human implantation for a period of greater than 29 days.
Product Details
Property Average Result
Durometer 24 Type A
Appearance Translucent
Elongation 800 %
Stress at Strain 320 psi / 300 % (2.21 Mpa / 300 %)
Tear 150 ppi (26.46 kN/m)
Tensile 1,200 psi (8.28 MPa)
Comment 100% solids