Implant Line
MED-6600
SILICONE DISPERSION
Description
  • Two-part, heat curable silicone dispersion
  • Cures via addition-cure chemistry
  • 1:1 Mix Ratio (Part A: Part B)
Applications
  • Suitable for dip casting and heat-curing of thin elastomeric films
  • Low viscosity makes dispersions ideal for use as sprayable coatings
NuSil Technology’s MED-6600 may be considered for use in human implantation for a period of greater than 29 days.
Product Details
Property Average Result
Cure System Platinum
Viscosity 350 cP (350 mPa*s)
Non Volatile Content 35 %
Durometer 25 Type A
Appearance Translucent
Cure 30 minutes / 25 °C 45 minutes / 75 °C 135 minutes / 150 °C
Elongation 800 %
Mix Ratio 1:1
Refractive Index 1.46
Specific Gravity 1.1
Stress at Strain 300 psi / 300 % (2.07 Mpa / 300 %)
Tear 160 ppi (28.22 kN/m)
Tensile 1,200 psi (8.28 MPa)
Work Time 24 hours
Comment 1.46 R.I. in Xylene