Description
- Two-part, low viscosity, heat curable silicone dispersion
- Cures via addition-cure chemistry
- 1:1 Mix Ratio (Part A: Part B)
Applications
- Suitable for dip casting and heat-curing of thin elastomeric films
- Low viscosity makes dispersions ideal for use as sprayable coatings
Product Details
Property | Average Result |
---|---|
Cure System | Platinum |
Viscosity | 860 cP (860 mPa*s) |
Non Volatile Content | 35 % |
Durometer | 30 Type A |
Appearance | Translucent |
Cure | 30 minutes / 25 °C 45 minutes / 75 °C 135 minutes / 150 °C |
Cure Type | Heat Cure |
Elongation | 850 % |
Mix Ratio | 1:1 |
Refractive Index | 1.43 |
Rheology | Flowable/Self Leveling |
Tear | 160 ppi (28.22 kN/m) |
Tensile | 1,600 psi (11.03 MPa) |