Description
- A two-part platinum, addition cured, low viscosity liquid that cures to a soft silicone gel suitable for applications requiring temporary adhesion.
- 100% silicone solids (solvent-less)
- 1:1 Mix Ratio (Part A: Part B)
- Supported with applicable biological testing
Applications
- Can be considered for use in wound care applications and scar care products
- Suitable for applications requiring a clear silicone gel with high adhesion
- Ideal for coating various substrates
Product Details
Property | Average Result |
---|---|
Penetration | 13 mm |
Viscosity | 8,500 cP (8,500 mPa*s) |
Appearance | Translucent |
Cure | 60 minutes / 140 °C |
Cure System | Platinum |
Mix Ratio | 1:1 |
Comment | Low Viscosity, Soft Gel |