Description
- A high purity, clear silicone gel manufactured under controlled conditions
- Resists fracturing under stress
- Easy mixing, two-component, low viscosity system
- 1:1 Mix Ratio (Part A: Part B)
- Alternate mix ratios may be considered to adjust penetration
Applications
- Ideal for backfilling and dampening
- Suitable for applications requiring a high purity, clear silicone gel that resists fracturing under stressed conditions
- NuSil Technology can formulate custom materials to meet specific penetration requirements
Product Details
Property | Average Result |
---|---|
Penetration | 2.6 mm |
Viscosity | 500 cP (500 mPa*s) |
Mix Ratio | 1:1 |
Appearance | Clear |
Cure | 3.5 hours / 160 °C |
Specific Gravity | 0.97 |
Volatile Content | 0.1 % |
Comment | Low penetration, high strength gel |