Implant Line
MED-6233
OPTICALLY CLEAR LOW VISCOSITY SILICONE ELASTOMER
Description
  • Pourable, optically clear, two-part silicone elastomer
  • Cures with heat via addition-cure chemistry
  • 1:1 Mix Ratio (Part A: Part B)
Applications
  • For use in potting, encapsulating, coating, and injection or transfer molding applications
NuSil Technology’s MED-6233 may be considered for use in human implantation for a period of greater than 29 days.
Product Details
Property Average Result
Durometer 50 Type A
Viscosity 73,500 cP (73,500 mPa*s)
Work Time 52 hours
Tensile 740 psi (5.1 MPa)
Appearance Clear
Cure 30 minutes / 150 °C
Cure System Platinum
Elongation 380 %
Mix Ratio 1:1
Refractive Index 1.41
Specific Gravity 1.04
Tear 80 ppi (14.11 kN/m)
Comment Clear, 1.41 R.I.