Description
- Flowable, two-part, clear, high durometer silicone
- Cures with heat via addition-cure chemistry
- 1:1 Mix Ratio (Part A: Part B)
Applications
- For use in potting, encapsulating, coating, and injection or transfer molding applications
- Useful in any application requiring a clear, flowable, high durometer material
Product Details
Property | Average Result |
---|---|
Durometer | 75 Type A |
Viscosity | 25,500 cP (25,500 mPa*s) |
Work Time | 2.5 hours |
Tensile | 1,300 psi (8.97 MPa) |
Appearance | Clear |
Cure | 2 hours / 50 °C |
Cure System | Platinum |
Elongation | 65 % |
Mix Ratio | 1:1 |
Specific Gravity | 1.07 |
Tear | 35 ppi (6.17 kN/m) |
Comment | Clear, 1.41 R.I., Low Viscosity, High Durometer |