Description
- Two-part, optically clear, solvent free, low viscosity silicone elastomer
- Cures with heat via addition-cure chemistry
- 10:1 Mix Ratio (Part A: Part B)
Applications
- For use in potting, encapsulating, coating, and injection or transfer molding applications
- Use in the casting of clear silicone parts
- Use as a coating, by means of dipping or painting.
Product Details
Property | Average Result |
---|---|
Durometer | 50 Type A |
Viscosity | 3,800 cP (3,800 mPa*s) |
Work Time | 5 hours |
Tensile | 1,250 psi (8.62 MPa) |
Appearance | Clear |
Cure | 15 minutes / 150 °C |
Cure System | Platinum |
Elongation | 100 % |
Mix Ratio | 10:1 |
Specific Gravity | 1.03 |
Tack Free Time | 16 hours |
Comment | Clear, 1.41 R.I. |