Description
- Optically clear, two-component silicone elastomer
- Cures with heat via addition-cure chemistry
- 1:1 Mix Ratio (Part A: Part B)
Applications
- For use in potting, encapsulating, coating, and injection or transfer molding applications
Product Details
Property | Average Result |
---|---|
Durometer | 50 Type A |
Viscosity | 16,000 cP (16,000 mPa*s) |
Work Time | 4 hours |
Tensile | 930 psi (6.41 MPa) |
Appearance | Translucent |
Cure | 30 minutes / 150 °C |
Cure System | Platinum |
Elongation | 140 % |
Mix Ratio | 1:1 |
Refractive Index | 1.43 |
Specific Gravity | 1.05 |
Tear | 35 ppi (6.17 kN/m) |
Comment | Clear, 1.43 R.I. |