Implant Line
MED-5870
LIQUID SILICONE RUBBER
Description
  • Two-part, translucent silicone system designed for use with injection molding equipment
  • Rapid cure with no post-cure required
  • 1:1 Mix Ratio (Part A: Part B)
Applications
  • For the injection molding of parts requiring a material with a high durometer including: molded rubber stoppers, gaskets, seals, valves, o-rings and other precision parts
  • Suitable for over-molding applications
  • Can be used with NuSil’s healthcare color masterbatches for applications requiring colored silicones
NuSil Technology’s MED-5870 may be considered for use in human implantation for a period of greater than 29 days.
Product Details
Property Average Result
Durometer 68 Type A
Tensile 1,250 psi (8.62 MPa)
Elongation 400 %
Tear 225 ppi (39.68 kN/m)
Appearance Translucent
Cure 5 minutes / 165 °C
Cure System Platinum
Extrusion Rate 100 g/minute
Specific Gravity 1.14
Stress at Strain 900 psi / 200 % (6.21 MPa / 200 %)
Work Time
Comment Injection Molding Elastomer