Description
- Two-part, translucent silicone system designed for use with injection molding equipment
- Rapid cure with no post-cure required
- 1:1 Mix Ratio (Part A: Part B)
Applications
- For the injection molding of parts requiring a material with a medium durometer including: molded rubber stoppers, gaskets, seals, valves, o-rings and other precision parts
- Suitable for over-molding applications
- Can be used with NuSil’s color masterbatches for applications requiring colored silicones
Product Details
Property | Average Result |
---|---|
Durometer | 40 Type A |
Tensile | 1,350 psi (9.31 MPa) |
Elongation | 680 % |
Tear | 160 ppi (28.22 kN/m) |
Appearance | Translucent |
Cure | 5 minutes / 165 °C |
Extrusion Rate | 115 g/minute |
Specific Gravity | 1.12 |
Stress at Strain | 300 psi / 200 % (2.07 Mpa / 200 %) |
Work Time | |
Comment | Injection Molding Elastomer |