Description
- Two-part, translucent silicone system designed for use with injection molding equipment
- Rapid cure with no post-cure required
- 1:1 Mix Ratio (Part A: Part B)
Applications
- For the injection molding of parts requiring a material with a medium durometer including: molded rubber stoppers, gaskets, seals, valves, o-rings and other precision parts
- Suitable for over-molding applications
- Can be used with NuSil’s healthcare color masterbatches for applications requiring colored silicones
Product Details
Property | Average Result |
---|---|
Durometer | 30 Type A |
Tensile | 1,340 psi (9.24 MPa) |
Elongation | 830 % |
Tear | 150 ppi (26.46 kN/m) |
Appearance | Translucent |
Cure | 5 minutes / 165 °C |
Cure System | Platinum |
Extrusion Rate | 145 g/minute |
Specific Gravity | 1.11 |
Stress at Strain | 160 psi / 200 % (1.1 MPa / 200 %) |
Work Time | |
Comment | Injection Molding Elastomer |