Description
- Two-part, translucent silicone system used with injection molding equipment
- Rapid cure with no post-cure required
- Cures with heat via addition-cure chemistry
- 1:1 Mix Ratio (Part A: Part B)
Applications
- For the injection molding of parts requiring a material with a high durometer including: molded rubber stoppers, gaskets, seals, valves, o-rings and other precision parts
- Suitable for over-molding applications
- Can be used with NuSil’s Healthcare color masterbatches for applications requiring colored silicones
Product Details
Property | Average Result |
---|---|
Durometer | 80 Type A |
Tensile | 1,000 psi (6.9 MPa) |
Elongation | 250 % |
Tear | 90 ppi (15.87 kN/m) |
Appearance | Translucent |
Cure | 5 minutes / 165 °C |
Cure Rate Scorch | 0.95 minutes |
Cure Rate T90 | 2.05 minutes |
Cure System | Platinum |
Extrusion Rate | 107.5 g/minute |
Mix Ratio | 1:1 |
Specific Gravity | 1.17 |
Stress at Strain | 650 psi / 100 % (4.48 MPa / 100 %) |
Work Time | |
Comment | Injection Molding Elastomer |